Thermo Scientific™ National Assembled Certified Vial Kits
Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.
Manufacturer:  Thermo Scientific™ CERT401587ASM
 View more versions of this product
Catalog No. 03-377-403
Specifications
Description
Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.
  • Fully lot tested including HPLC and GC analysis
  • Certificate of Conformance included in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
  • Assembled in convenient packs of 100 vials and pre-attached caps and septa
Description & Specifications
Specifications
limited-volume sampling
13-425mm
Gray
Screw Thread
4mL Amber Glass Vial Kit, 13-425 Screw Thread
Pre-slit
Assembled Vial Kit
4mL
Bonded PTFE/Silicone, Preslit
4mL Amber Vial Kit, 13-425 Screw Thread
Bonded Red PTFE/White Silicone
Amber