Please login to your online account to display your discounted pricing

Nateglinide, Tocris™

Kir 6 (KATP ) blocker; displays high affinity for SUR1/Kir 6.2 channels

$142.08 - $593.12

Chemical Identifiers

Chemical Name or Material Nateglinide
Assay Percent Range >98%
Molecular Formula C19H27NO3
Formula Weight 317.42
Synonym N-[[4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423110

 
r & d systems
RDS423110
10mg Each for $142.08

423150

 
r & d systems
RDS423150
50mg Each for $593.12
Description & Specifications

Specifications

Formula Weight 317.42
Quantity 10mg
Chemical Name or Material Nateglinide
Assay Percent Range >98%
Molecular Formula C19H27NO3
CAS 105816-04-4
Synonym N-[[4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature