Nasco Sampling Whirl-Pak™ Easy-To-Close Bags
Features puncture proof tabs for protection from damage due to wire-end protrusion. Whirl-Pak™ Easy-To-Close Bags are ideal for sampling and product analysis.
Manufacturer:  Nasco Sampling B01186WAFB
Catalog No. 01-815-22
Description
Description
  • Transparent polyethylene bag
  • Gas sterilized, with documentation available
  • Pull tabs help prevent contamination from finger contact when opening
  • Write-on bags have area that accepts ballpoint pen or pencil
  • Bag can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing 
  • Thickness, 0.05mm
Bags should not be used at temperatures above 180°F (82°C).
Specifications
Specifications
Write-on Style
532mL
0.05mm
23 x 11.5cm
PE (Polyethylene)