missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nasco Whirl-Pak™ Standard Sample Bags
Features puncture proof tabs for protection from damage due to wire-end protrusion. Nasco™ Whirl-Pak™ Easy-To-Close Bags are ideal for sampling and product analysis.
Manufacturer: Nasco B01195WA
Description
Features puncture proof tabs for protection from damage due to wire-end protrusion. Nasco™ Whirl-Pak™ Easy-To-Close Bags are ideal for sampling and product analysis.
- Transparent polyethylene bags
- Gas sterilized, with documentation available
- Pull tabs help prevent contamination from finger contact when opening
- Write-on bags have area that accepts ballpoint pen or pencil
- Bags can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing
- Thickness, 2.25 to 4mil
Bags should not be used at temperatures above 180°F (82°C).
Specifications
Write-on Style | |
4 mil | |
1.63 L | |
Transparent | |
PE (Polyethylene) | |
19 cm |
30 x 19cm | |
55 oz. | |
Closing Tab | |
30 cm | |
0.102mm |