Nasco Sludge Judge™ Liquid Samplers
Made from rigid plastic tubing in three 5 ft. (1.53m) sections, provides maximum strength for liquid sampling at long distances
$241.79 - $378.23
Specifications
Certifications/Compliance ISO 9001:2008 and ISO 13485:2003
For Use With (Application) Sampling 5% or less solids
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Outer Tube Length (English) Material Product Type Price Quantity  
Catalog Number Mfr. No. Diameter (English) Outer Tube Length (English) Material Product Type Price Quantity  
14-209-140 Nasco
C09247WA
0.75 in. Overall: 15 ft.; Section: 5 ft. Polycarbonate Sludge Judge™ Sampler Each for $241.79
01-812-79 Nasco
B01355WA
1.25 in. Overall: 15 ft.; Section: 5 ft. Polycarbonate with Ultraviolet Stabilizer Sludge JudgeS II Large Capacity Sampler Each for $378.23
01-812-95 Nasco
B01386WA
0.75 in. Overall: 15 ft.; Section: 5 ft. Polycarbonate Sludge Judge™ Ultra Sampler Each for $353.77
Specifications
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
ISO 9001:2008 and ISO 13485:2003
Sampling 5% or less solids