Please login to your online account to display your discounted pricing

NAS-181, Tocris

Selective r5-HT1B antagonist; Active in vivo

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material NAS-181
Assay Percent Range >97%
Molecular Formula C19H26N2O4·2CH3SO3H
Formula Weight 538.63
Synonym (2R)-2-[[[3-(4-Morpholinylmethyl)-2H-1-benzopyran-8-yl]oxy]methyl]morpholine dimethanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

141310

 
r & d systems
141310
10mg Each for $284.81

141350

 
r & d systems
141350
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 538.63
Quantity 10mg
Chemical Name or Material NAS-181
Assay Percent Range >97%
CAS 1217474-40-2
Molecular Formula C19H26N2O4·2CH3SO3H
Synonym (2R)-2-[[[3-(4-Morpholinylmethyl)-2H-1-benzopyran-8-yl]oxy]methyl]morpholine dimethanesulfonate
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C