missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naringin hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1016318

 View more versions of this product

Catalog No. AAL1016318



Description

Description

A citrus bioflavanoid that inhibits cytochrome P450 monooxygenase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Naringin hydrate
∽166°C (decomposition)
MFCD00148888,MFCD00149445,MFCD01461988
102012
DFPMSGMNTNDNHN-ZPHOTFPESA-N
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
580.54
580.55 (Anhydrous)
132203-74-8
C27H32O14
50 g
14,6424
−90° (c=1 in Ethanol)
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
74787988
98%

Chemical Identifiers

132203-74-8
580.54
DFPMSGMNTNDNHN-ZPHOTFPESA-N
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C27H32O14
MFCD00148888,MFCD00149445,MFCD01461988
74787988
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
Safety and Handling

Safety and Handling

EINECSNumber : 233-566-4

RTECSNumber : QN6340000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only