Please login to your online account to display your discounted pricing

Naringin, 97%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC206911000

 View more versions of this product

Catalog No. AC206911000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 580.53
Quantity 100g
Color Beige to yellow
Specific Rotation Condition −91° (20°C c=1,C2H5OH)
Merck Index 15, 6507
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Naringin
Assay Percent Range 97%
Molecular Formula C27H32O14
CAS 10236-47-2
Specific Rotation -89° to -93° (20°C, 589nm) (c=1, C2H5OH)
MDL Number MFCD00148888
Infrared Spectrum Authentic
Packaging Plastic bottle

Specifications

Chemical Name or Material Naringin
CAS 10236-47-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C27H32O14
Formula Weight 580.53
MDL Number MFCD00148888