missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Naringin, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 206910250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Naringin | |
96.0 | |
97% | |
MFCD00148888,MFCD00149445,MFCD01461988 | |
DFPMSGMNTNDNHN-ZPHOTFPESA-N | |
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O | |
580.54 | |
580.53 | |
Plastic bottle | |
25g |
10236-47-2 | |
100.0 | |
C27H32O14 | |
15, 6507 | |
Authentic | |
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one | |
74787988 | |
≥96.0% | |
Beige to Yellow |
Chemical Identifiers
10236-47-2 | |
580.54 | |
DFPMSGMNTNDNHN-ZPHOTFPESA-N | |
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one |
C27H32O14 | |
MFCD00148888,MFCD00149445,MFCD01461988 | |
74787988 | |
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O |
RUO â Research Use Only