Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthol Yellow S

$40.69 - $513.81

Chemical Identifiers

Chemical Name or Material Naphthol Yellow S
CAS 846-70-8
Molecular Formula C10H4N2Na2O8S
Formula Weight 358.2
MDL Number MFCD00003958
Beilstein 2708476
Merck Index 14,6393
Synonym C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB208720B

  SDS
alfa aesar™
B208720B
1000g Each for $513.81

AAB2087218

  SDS
alfa aesar™
B2087218
50g Each for $40.69

AAB2087230

  SDS
alfa aesar™
B2087230
250g Each for $159.72
Description & Specifications

Specifications

Formula Weight 358.2
Molecular Formula C10H4N2Na2O8S
Melting Point >300°C
Chemical Name or Material Naphthol Yellow S
MDL Number MFCD00003958
Beilstein 2708476
RTECS Number QK1813000
Synonym C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
TSCA TSCA
Merck Index 14,6393
Quantity 1000g
CAS 846-70-8