missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthenic acids, pract., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 415285000

 View more versions of this product

Catalog No. AC415285000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Naphthenic acids
230 to 260mg KOH/g
0.9100g/mL
149°C
Glass bottle
500 g
Solubility in water: insoluble
Liquid
pract.
1338-24-5
140.0°C to 200.0°C
Authentic
MFCD00147696
0.91
5 stokes max. (25°C)

Chemical Identifiers

1338-24-5
MFCD00147696
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 215-662-8

RTECSNumber : QK8750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only