Please login to your online account to display your discounted pricing

Naphthalene-d8, for NMR, +98% atom D, ACROS Organics™

$71.47 - $71.47

Chemical Identifiers

Chemical Name or Material Naphthalene-d8
Name Note For NMR
CAS 1146-65-2
Purity Grade For NMR
Molecular Formula C10D8
Linear Formula C10D8
Formula Weight 136.22
MDL Number MFCD00001743
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC174960050

  SDS
thermo fisher scientific
AC174960050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC174960010

  SDS
thermo fisher scientific
174960010
Glass bottle 1g Each for $71.47
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Formula Weight 136.22
Quantity 5g
Color White
Purity Grade For NMR
Chemical Name or Material Naphthalene-d8
Molecular Formula C10D8
Linear Formula C10D8
Name Note For NMR
MDL Number MFCD00001743
CAS 1146-65-2
Deuterium (D) 98.0% min.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For NMR

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.