missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene (Crystalline/Laboratory), Fisher Chemical

Manufacturer:  Fisher Chemical N7500

Catalog No. N7-500




Description

Description

Specifications

Specifications

Naphthalene
Pass Test
0.08mbar at 20°C
White
500g
C10H8
albocarbon, camphor tar, moth balls, moth flakes, naphtalene, naphthalin, naphthaline, naphthene, tar camphor, white tar
C1=CC2=CC=CC=C2C=C1
128.17
CHEBI:16482
Solid
0.990g/cm³
Poly Bottle
218°C
79°C
91-20-3
MFCD00001742
UFWIBTONFRDIAS-UHFFFAOYSA-N
naphthalene
931
128.18
Laboratory

Chemical Identifiers

91-20-3
128.17
UFWIBTONFRDIAS-UHFFFAOYSA-N
931
naphthalene
C10H8
MFCD00001742
albocarbon, camphor tar, moth balls, moth flakes, naphtalene, naphthalin, naphthaline, naphthene, tar camphor, white tar
CHEBI:16482
C1=CC2=CC=CC=C2C=C1
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable solid. Possible cancer hazard May cause cancer based on animal data. May cause skin, eye, and respiratory tract irritation. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0

SDS Documents
Product Certifications