Learn More
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180202500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopySpecifications
218.0°C | |
79.0°C to 82.0°C | |
Powder | |
99.0 | |
0.9900g/mL | |
250 g | |
99+% | |
C10H8 | |
MFCD00001742 | |
15, 6455 | |
Solubility in water: 30 mg/l (25°C). Other solubilities: soluble in ethanol,, very soluble in acetone, chloroform, ether,, benzene, toluene, carbon disulfide | |
C1=CC2=CC=CC=C2C=C1 | |
128.17 | |
CHEBI:16482 | |
≥99% |
Naphthalene | |
99+% | |
91-20-3 | |
100.0 | |
Beige to Red/Salmon | |
78°C | |
Authentic | |
C10H8 | |
05, IV, 1640 | |
Glass Bottle | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
naphthalene | |
931 | |
128.17 | |
0.99 |
Chemical Identifiers
91-20-3 | |
128.17 | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
CHEBI:16482 | |
C1=CC2=CC=CC=C2C=C1 |
C10H8 | |
MFCD00001742 | |
931 | |
naphthalene |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-049-5
RTECSNumber : QJ0525000
TSCA : TSCA
RUO – Research Use Only