missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 180200010

 View more versions of this product

Catalog No. AC180200010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For fluorescence/luminescence spectroscopy
Specifications

Specifications

218.0°C
79.0°C to 82.0°C
Powder
99.0
0.9900g/mL
1 kg
99+%
C10H8
MFCD00001742
15, 6455
Solubility in water: 30 mg/l (25°C). Other solubilities: soluble in ethanol,, very soluble in acetone, chloroform, ether,, benzene, toluene, carbon disulfide
C1=CC2=CC=CC=C2C=C1
128.17
CHEBI:16482
≥99%
Naphthalene
99+%
91-20-3
100.0
Beige to Red/Salmon
78°C
Authentic
C10H8
05, IV, 1640
Glass Bottle
UFWIBTONFRDIAS-UHFFFAOYSA-N
naphthalene
931
128.17
0.99

Chemical Identifiers

91-20-3
128.17
UFWIBTONFRDIAS-UHFFFAOYSA-N
CHEBI:16482
C1=CC2=CC=CC=C2C=C1
C10H8
MFCD00001742
931
naphthalene
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-049-5

RTECSNumber : QJ0525000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only