Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-2-sulfonic acid hydrate, 94%

Chemical Identifiers

Chemical Name or Material Naphthalene-2-sulfonic acid hydrate
CAS 76530-12-6
Molecular Formula C10H8O3S·xH2O
Formula Weight 208.23 (Anhydrous)
MDL Number MFCD00149271
UN Number UN2585
Merck Index 14,6377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA134450B

  SDS
alfa aesar™
A134450B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1344518

  SDS
alfa aesar™
A1344518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1344530

  SDS
alfa aesar™
A1344530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.23 (Anhydrous)
Chemical Name or Material Naphthalene-2-sulfonic acid hydrate
TSCA TSCA
Assay 94%
Molecular Formula C10H8O3S·xH2O
MDL Number MFCD00149271
CAS 76530-12-6
UN Number UN2585
Merck Index 14,6377
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.