Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-2-sulfonamide, 97%

$101.09 - $361.62

Chemical Identifiers

Chemical Name or Material Naphthalene-2-sulfonamide
CAS 1576-47-2
Molecular Formula C10H9NO2S
Formula Weight 207.25
MDL Number MFCD00051931
Beilstein 1955730
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0916406

  SDS
alfa aesar™
L0916406
5g Each for $101.09

AAL0916414

  SDS
alfa aesar™
L0916414
25g Each for $361.62
Description & Specifications

Specifications

Formula Weight 207.25
Chemical Name or Material Naphthalene-2-sulfonamide
MDL Number MFCD00051931
Molecular Formula C10H9NO2S
Assay 97%
Quantity 5g
Melting Point 215° to 222°C
CAS 1576-47-2
Beilstein 1955730