Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-2-carboxamide, 98%

$108.97 - $423.30

Chemical Identifiers

Chemical Name or Material Naphthalene-2-carboxamide
CAS 2243-82-5
Molecular Formula C11H9NO
Formula Weight 171.2
MDL Number MFCD00016812
Beilstein 1101108
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0839104

  SDS
alfa aesar™
L0839104
2g Each for $108.97

AAL0839109

  SDS
alfa aesar™
L0839109
10g Each for $423.30
Description & Specifications

Specifications

Formula Weight 171.2
Chemical Name or Material Naphthalene-2-carboxamide
Assay 98%
Molecular Formula C11H9NO
MDL Number MFCD00016812
CAS 2243-82-5
Beilstein 1101108
Melting Point 193° to 197°C
Quantity 2g