Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-2-carbonitrile, 97%

Chemical Identifiers

Chemical Name or Material Naphthalene-2-carbonitrile
CAS 613-46-7
Molecular Formula C11H7N
Formula Weight 153.18
MDL Number MFCD00016807
UN Number UN3439
Beilstein 1363868
Synonym 2-Cyanonaphthalene; 2-Naphthonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0370406

 
alfa aesar™
L0370406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0370414

 
alfa aesar™
L0370414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.18
Chemical Name or Material Naphthalene-2-carbonitrile
Assay 97%
MDL Number MFCD00016807
Synonym 2-Cyanonaphthalene; 2-Naphthonitrile
Health Hazard 2 GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Molecular Formula C11H7N
CAS 613-46-7
UN Number UN3439
Beilstein 1363868
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number QL5976350
Boiling Point 156° to 158°C (12mmHg)
Melting Point 63° to 67°C
Quantity 5g
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.