missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Naphthalene-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6467103
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Naphthalene-2-boronic acid pinacol ester | |
C16H19BO2 | |
1 g | |
Slightly soluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2 | |
254.136 | |
254.14 | |
Solid |
256652-04-7 | |
MFCD05663868 | |
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668 | |
SPPZBAGKKBHZRW-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane | |
10753409 | |
97% |
Chemical Identifiers
256652-04-7 | |
254.136 | |
SPPZBAGKKBHZRW-UHFFFAOYSA-N | |
10753409 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2 |
C16H19BO2 | |
MFCD05663868 | |
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane, naphthalene-2-boronic acid pinacol ester, naphthalene-2-boronic acid, pinacol ester, 2-naphthaleneboronic acid, pinacol ester, 2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl, 2-naphthaleneboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane, amtb668 | |
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only