Learn More
Naphthalene-1-sulfonic acid sodium salt, 98% (dry wt.), water <3.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2248822
Description
Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNaphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Naphthalene-1-sulfonic acid sodium salt | |
299°C to 301°C | |
C10H7NaO3S | |
100 g | |
Hygroscopic | |
Soluble in water. | |
C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+] | |
230.213 | |
230.22 |
130-14-3 | |
(dry wt.), water <2% | |
MFCD00064964 | |
3639038 | |
sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn | |
HIEHAIZHJZLEPQ-UHFFFAOYSA-M | |
sodium;naphthalene-1-sulfonate | |
23661867 | |
99% |
Chemical Identifiers
130-14-3 | |
230.213 | |
HIEHAIZHJZLEPQ-UHFFFAOYSA-M | |
23661867 | |
C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+] |
C10H7NaO3S | |
MFCD00064964 | |
sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn | |
sodium;naphthalene-1-sulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 204-976-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only