missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Naphthalene-1-sulfonic acid hydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2178218

 View more versions of this product

Catalog No. AAB2178218



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Naphthalene-1-sulfonic acid hydrate
89°C to 91°C (dried)
MFCD00003987
UN2585
naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
208.23
208.23 (Anhydrous)
6036-48-2
C10H8O3S
50 g
14,6376
PSZYNBSKGUBXEH-UHFFFAOYSA-N
naphthalene-1-sulfonic acid;dihydrate
71431272
98%

Chemical Identifiers

6036-48-2
208.23
PSZYNBSKGUBXEH-UHFFFAOYSA-N
71431272
OS(=O)(=O)C1=C2C=CC=CC2=CC=C1
C10H8O3S
MFCD00003987
naphthalene-1-sulfonic acid dihydrate, 1-naphthalenesulfonic acid dihydrate, naphthalene-1-sulfonic acid hydrate
naphthalene-1-sulfonic acid;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

EINECSNumber : 201-610-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only