Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-1-carboxamide, 98%

$48.34 - $152.39

Chemical Identifiers

Chemical Name or Material Naphthalene-1-carboxamide
CAS 2243-81-4
Molecular Formula C11H9NO
Formula Weight 171.2
MDL Number MFCD00014314
Beilstein 2044731
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0836403

 
alfa aesar™
L0836403
1g Each for $48.34

AAL0836406

 
alfa aesar™
L0836406
5g Each for $152.39
Description & Specifications

Specifications

Formula Weight 171.2
Chemical Name or Material Naphthalene-1-carboxamide
Assay 98%
Molecular Formula C11H9NO
MDL Number MFCD00014314
CAS 2243-81-4
Beilstein 2044731
Melting Point 206° to 210°C
Quantity 1g