Learn More
Naphthalene-1-carbonitrile, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1449814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Naphthalene-1-carbonitrile | |
33°C to 38°C | |
C11H7N | |
25 g | |
971255 | |
YJMNOKOLADGBKA-UHFFFAOYSA-N | |
naphthalene-1-carbonitrile | |
6846 | |
95% |
86-53-3 | |
299°C | |
MFCD00003866 | |
UN3439 | |
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd | |
C1=CC=C2C(=C1)C=CC=C2C#N | |
153.184 | |
153.18 |
Chemical Identifiers
86-53-3 | |
153.184 | |
YJMNOKOLADGBKA-UHFFFAOYSA-N | |
6846 | |
C1=CC=C2C(=C1)C=CC=C2C#N |
C11H7N | |
MFCD00003866 | |
1-naphthonitrile, 1-cyanonaphthalene, 1-naphthalenecarbonitrile, naphthalenecarbonitrile, 1-naphthylnitrile, alpha-naphthonitrile, 1-naphthalenenitrile, alpha-naphthylnitrile, alpha-cyanonaphthalene, unii-2as1lz61hd | |
naphthalene-1-carbonitrile |
Safety and Handling
GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 201-679-8
RTECSNumber : QL5976300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only