missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-1,4-dithiocarboxamide, 97%

$33.86 - $439.73

Chemical Identifiers

CAS 1347815-21-7
Molecular Formula C12H10N2S2
Molecular Weight (g/mol) 246.346
MDL Number MFCD20265223
InChI Key RMVWFHNLLYPEGN-UHFFFAOYSA-N
Synonym naphthalene-1,4-bis carbothioamide
PubChem CID 73995732
IUPAC Name naphthalene-1,4-dicarbothioamide
SMILES C1=CC=C2C(=C1)C(=CC=C2C(=S)N)C(=S)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54278MD View Documents Alfa Aesar™
H54278MD
250mg Each for $33.86
 
AAH5427803 View Documents Alfa Aesar™
H5427803
1g Each for $106.09
 
AAH5427806 View Documents Alfa Aesar™
H5427806
5g Each for $439.73
 
Specifications

Chemical Identifiers

1347815-21-7
246.346
RMVWFHNLLYPEGN-UHFFFAOYSA-N
73995732
C1=CC=C2C(=C1)C(=CC=C2C(=S)N)C(=S)N
C12H10N2S2
MFCD20265223
naphthalene-1,4-bis carbothioamide
naphthalene-1,4-dicarbothioamide

Specifications

250mg
C12H10N2S2
naphthalene-1,4-bis carbothioamide
C1=CC=C2C(=C1)C(=CC=C2C(=S)N)C(=S)N
246.346
246.35
Naphthalene-1,4-dithiocarboxamide
1347815-21-7
MFCD20265223
RMVWFHNLLYPEGN-UHFFFAOYSA-N
naphthalene-1,4-dicarbothioamide
73995732
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures