missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Naphthalene-1,4-dithiocarboxamide, 97%

Manufacturer:  Alfa Aesar™ H5427806

 View more versions of this product

Catalog No. AAH5427806



Specifications

Specifications

Naphthalene-1,4-dithiocarboxamide
1347815-21-7
MFCD20265223
RMVWFHNLLYPEGN-UHFFFAOYSA-N
naphthalene-1,4-dicarbothioamide
73995732
97%
5g
C12H10N2S2
naphthalene-1,4-bis carbothioamide
C1=CC=C2C(=C1)C(=CC=C2C(=S)N)C(=S)N
246.346
246.35

Chemical Identifiers

1347815-21-7
246.346
RMVWFHNLLYPEGN-UHFFFAOYSA-N
73995732
C1=CC=C2C(=C1)C(=CC=C2C(=S)N)C(=S)N
C12H10N2S2
MFCD20265223
naphthalene-1,4-bis carbothioamide
naphthalene-1,4-dicarbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents