missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Naphthalene-1,4-dicarbonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2024814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Naphthalene-1,4-dicarbonitrile | |
208°C to 212°C | |
C12H6N2 | |
25 g | |
1,4-dicyanonaphthalene, 1,4-naphthalenedicarbonitrile, 1,4-dicyanonahthalene, acmc-209heb, ghl.pd_mitscher_leg0.246 | |
C1=CC=C2C(=CC=C(C2=C1)C#N)C#N | |
178.194 | |
178.19 |
3029-30-9 | |
Odorless | |
MFCD03093626 | |
1870226 | |
BENSWQOUPJQWMU-UHFFFAOYSA-N | |
naphthalene-1,4-dicarbonitrile | |
76414 | |
≥98% |
Chemical Identifiers
3029-30-9 | |
178.194 | |
BENSWQOUPJQWMU-UHFFFAOYSA-N | |
76414 | |
C1=CC=C2C(=CC=C(C2=C1)C#N)C#N |
C12H6N2 | |
MFCD03093626 | |
1,4-dicyanonaphthalene, 1,4-naphthalenedicarbonitrile, 1,4-dicyanonahthalene, acmc-209heb, ghl.pd_mitscher_leg0.246 | |
naphthalene-1,4-dicarbonitrile |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P280f
Wear protective clothing.
Warning
EINECSNumber : 221-197-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only