missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ NanoDrop™ 8000 Spectrophotometer

Thermo Scientific™ NanoDrop 8000 is a muti-sample micro-volume UV-Vis spectrophotometer.

Manufacturer:  Thermo Scientific™ ND8000USCAN

 View more versions of this product

Catalog No. ND8000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disclaimers:

US Patents 6,628,382 and 6,809,826.


Includes: USB cable and operating software

Specifications

Description

Thermo Scientific™ NanoDrop 8000 is a muti-sample micro-volume UV-Vis spectrophotometer. With the NanoDrop 8000, you can measure more samples in less time with the same reliability and patented sample retention technology of the NanoDrop 2000.

The NanoDrop 8000 allows scientists to quickly and easily quantify and assess purity of samples such as proteins and nucleic acids. Using an 8-channel pipetter to dispense samples onto an array of pedestals, analyze 96 samples in less than six minutes.

Key Features and Benefits
  • Designed for micro-volume (1μL) samples
  • No dilutions required, even for high concentration samples
  • Direct, easy measurements from tubes or 96-well plates
  • Can measure multiple samples in one measurement cycle, up to eight samples at one time
  • Single sample mode
  • Preconfigured methods for common applications such as Nucleic Acid, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, Lowry and more
  • User-friendly software for creating custom methods and options to design reports and export data
  • Patented sample retention system eliminates the need for cuvets
  • Full spectrum (220 to 750nm)
  • Calculates sample purity ratios (260/280)

Compatible with:

Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, and Vista™ (32-bit).

Description & Specifications

Specifications

3% (at 0.74 Abs at 350nm)
0 - 75Abs
3% (at .74 at 350nm)
UL/CSA and CE
USB
2.5 ng/µL (dsDNA), 0.15 mg/mL (BSA)
2048-element linear silicon CCD array
USB cable and operating software
Xenon Flash Lamp
Up to 8
12VDC
≤3 nm (FWHM at Hg 546.1 nm)
Spectrophotometer
±1 nm
220 - 750 nm
1 year parts and labor
0.003 *SD of 10 individual measurements at 0.74Abs
Nucleic acid, protein, cell culture and custom methods
Professional versions of Microsoft™ Windows™ 7 (32 bit and 64 bit), Windows Vista (32 bit), and Windows XP (32 bit).
NanoDrop 8000 Microvolume UV-Vis spectrophotometer
2.5-3,700 ng/µL (dsDNA), 0.15 - 100 mg/mL (BSA)
24 x 32 cm
ND-8000
<20 sec.
0.1, 0.2 and 1.0 mm
1µL
Microsoft Windows XP (32-bit) w/Service Pack (SP) 2 or later,Vista (32-bit), and Windows 7 (32 bit and 64 bit)
12VDC
±1 nm
3.4 kg