missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ NanoDrop™ 2000/2000c Spectrophotometers

Thermo Scientific™ NanoDrop 2000 and 2000c are full-spectrum, UV-Vis spectrophotometers used to quantify and assess purity of DNA, RNA, Protein and more.

Manufacturer:  Thermo Scientific™ ND2000USCAN

 View more versions of this product

Catalog No. ND2000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Thermo Scientific™ NanoDrop 2000 and 2000c are full-spectrum, UV-Vis spectrophotometers used to quantify and assess purity of DNA, RNA, Protein and more. The NanoDrop 2000 and 2000c are the only microvolume spectrophotometers with patented sample retention technology that measure sample volumes as small as 0.5μL.

The patented sample retention system of the NanoDrop 2000 and 2000c spectrophotometers allows a sample to be pipetted directly onto an optical measurement surface. Once the measurement is complete, the surfaces are simply wiped with a lint-free lab wipe. The NanoDrop 2000c integrates the sample retention system with cuvette capability.

NanoDrop 2000

  • Micro-volume sample size (0.5-2.0μL)
  • Easy to use—pipet sample directly onto the pedestal
  • No dilutions required, even for highly concentrated samples
  • Fast measurement time of less than 5 seconds
  • Full spectral output (190 - 840 nm)
  • Preconfigured methods for A260, A280, labeled DNA or Proteins, and much more
  • User-friendly software that includes custom methods and data export capabilities

NanoDrop 2000c

  • Combines all features of the NanoDrop 2000 with cuvette capability in one instrument
  • Advanced micro-volume pedestal technology
  • Dual modes—choose cuvette or pedestal
  • Broader concentration range for measuring very low or very high concentrations
  • Cuvette capability allows for kinetics (time or time/temperature studies) and cell culture (OD 600) measurements

NanoDrop 2000c - Cuvette

  • Heating: 37°C, ±0.5°C
  • Stirring: 150 - 850rpm
  • Z-Height: 8.5mm
  • Cuvette dimensions: 12.5 x 12.5mm, up to 48mmH
  • Cuvette path length options: 10, 5, 2 and 1mm
  • Type: Masked cuvette
  • Absorbance Range: 0.008 to 1.5
  • Measurement Time: <3 sec.
  • Weight: 2.1kg

Compatible with:

Professional versions of Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, Vista™ (32-bit), and Windows™ 7 (32 bit and 64 bit)

Specifications

Specifications

3% (at 0.74 Abs at 350 nm)
0.002 *SD of 10 individual measurements at 0.74 Abs
UL/CSA and CE
Microvolume Spectrophotometer
2048-element linear silicon CCD array
---
Xenon flash lamp
1
≤1.8nm (FWHM at Hg 253.7)
Spectrophotometer
5W
190 - 840 nm
One year parts and labor
0 - 300 Abs
Nucleic acid, protein, cell culture and custom methods
USB
2-15,000ng/µL (dsDNA), 0.10 - 400mg/mL (BSA)
14 x 20cm
NanoDrop 2000
<5 sec.
0.5-2.0µL
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
12VDC
±1nm
2.0 kg
Documents

US Patents 6,628,382 and 6,809,826.