Promega Nano-Glo Live Cell Assay System DFS Item
The Nano-Glo™ Live Cell Assay System is a single-addition, non-lytic detection reagent used to measure NanoBiT or NanoLuc luminescence from living cells.
Supplier:  Promega N2012
 View more versions of this product
Catalog No. PRN2012
Description
Description
  • Measures NanoLuc(R) Luminescence in Living Cells
  • Substrate and buffer optimized to enhance stability
  • Use to detect NanoBiT(R) protein complementation or NanoLuc(R) reporter activity
  • Monitors luminescence at a single time point or continuously for up to 2 hours without compromising cell viability.
Specifications
Specifications
-30°C to -10°C
NanoBiT™ protein complementation or NanoLuc™ reporter activity
Substrate and Buffer
1000 Assays