Please login to your online account to display your discounted pricing

NAN-190 hydrobromide, Tocris

5-HT1A antagonist

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material NAN-190 hydrobromide
Assay Percent Range >98%
Molecular Formula C23H27N3O3·HBr
Formula Weight 474.4
Synonym 1-(2-Methoxyphenyl)-4-(4-phthalimidobutyl)piperazine hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

0553250

 
r & d systems
0553250
50mg Each for $534.65

055350

 
r & d systems
055350
50mg Each for $126.32
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 474.4
Chemical Name or Material NAN-190 hydrobromide
Assay Percent Range >98%
Molecular Formula C23H27N3O3·HBr
CAS 115338-32-4
Synonym 1-(2-Methoxyphenyl)-4-(4-phthalimidobutyl)piperazine hydrobromide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature