missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NAM 98.0+%, TCI America™

Manufacturer:  TCI America A5591100MG

 View more versions of this product

Catalog No. A5591100MG



Specifications

Specifications

NAM [=N-(9-Acridinyl)maleimide] [for HPLC Labeling]
100mg
C17H10N2O2
N-(9-Acridinyl)maleimide, 9-Maleimidoacridine
C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N4C(=O)C=CC4=O
274.279
274.28
≥98.0% (HPLC)
Yellow
49759-20-8
MFCD00042983
NCFIKBMPEOEIED-UHFFFAOYSA-N
1-acridin-9-ylpyrrole-2,5-dione
3016496
Crystal-Powder at 20°C

Chemical Identifiers

49759-20-8
274.279
NCFIKBMPEOEIED-UHFFFAOYSA-N
3016496
C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N4C(=O)C=CC4=O
C17H10N2O2
MFCD00042983
N-(9-Acridinyl)maleimide, 9-Maleimidoacridine
1-acridin-9-ylpyrrole-2,5-dione
Safety and Handling

Safety and Handling

TSCA : No

Documents