Please login to your online account to display your discounted pricing

Naltrindole hydrochloride, Tocris

δ selective non-peptide antagonist

$175.83 - $727.58

Chemical Identifiers

Chemical Name or Material Naltrindole hydrochloride
Assay Percent Range >99%
Molecular Formula C26H26N2O3·HCl
Formula Weight 450.96
Synonym 17-(Cyclopropylmethyl)-6,7-dehydro-4,5α-epoxy-3,14-dihydroxy-6,7-2',3'-indolomorphinan hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

074010

 
r & d systems
RDS074010
10mg Each for $175.83

074050

 
r & d systems
RDS074050
50mg Each for $727.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 450.96
CAS 111469-81-9
Molecular Formula C26H26N2O3·HCl
Chemical Name or Material Naltrindole hydrochloride
Synonym 17-(Cyclopropylmethyl)-6,7-dehydro-4,5α-epoxy-3,14-dihydroxy-6,7-2',3'-indolomorphinan hydrochloride
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature