Please login to your online account to display your discounted pricing

Naltriben mesylate, Tocris

Standard Glc deltasubscript 2 selective antagonist

$195.49 - $816.90

Products
Catalog Number Mfr. No. Quantity Price Quantity    

089250

 
r & d systems
RDS089250
50mg Each for $816.90

089210

 
r & d systems
RDS089210
10mg Each for $195.49
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 511.59
Recommended Storage Desiccate at Room Temperature
Synonym 17-(Cyclopropylmethyl)-6,7-didehydro-3,14β-dihydroxy-4,5α-epoxy-6,7-2',3'-benzo[b]furanomorphinan mesylate
Chemical Name or Material Naltriben mesylate
Molecular Formula C26H25NO4·CH3SO3H
CAS 122517-78-6
Assay Percent Range >98%
Safety and Handling

Desiccate at Room Temperature