missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha-(tert-Butoxycarbonyl)-tele-(p-toluenesulfonyl)-L-histidine, TCI America™

$131.39 - $456.81

Chemical Identifiers

CAS 35899-43-5
Molecular Formula C18H23N3O6S
Molecular Weight (g/mol) 409.457
MDL Number MFCD00065967
InChI Key DCLJSEPKYJSEHW-HNNXBMFYSA-N
Synonym boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
PubChem CID 6455010
IUPAC Name (2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20665G View Documents TCI America
B20665G
5g Each for $131.39
 
B206625G View Documents TCI America
B206625G
25g Each for $456.81
 
Specifications

Chemical Identifiers

35899-43-5
409.457
DCLJSEPKYJSEHW-HNNXBMFYSA-N
6455010
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
C18H23N3O6S
MFCD00065967
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

White-Yellow
5g
C18H23N3O6S
boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
409.457
409.46
≥98.0% (T)
125°C
35899-43-5
MFCD00065967
DCLJSEPKYJSEHW-HNNXBMFYSA-N
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
6455010
Crystal-Powder at 20°C
Nalpha-(tert-Butoxycarbonyl)-tele-(p-toluenesulfonyl)-L-histidine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0933

TSCA : No