missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha-(tert-Butoxycarbonyl)-L-glutamine, TCI America™

$111.53 - $190.97

Chemical Identifiers

CAS 13726-85-7
Molecular Formula C10H18N2O5
Molecular Weight (g/mol) 246.263
MDL Number MFCD00065571
InChI Key VVNYDCGZZSTUBC-LURJTMIESA-N
Synonym boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine
PubChem CID 83687
IUPAC Name (2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B164910G View Documents TCI America
B164910G
10g Each for $111.53
 
B164925G View Documents TCI America
B164925G
25g Each for $190.97
 
Specifications

Chemical Identifiers

13726-85-7
246.263
VVNYDCGZZSTUBC-LURJTMIESA-N
83687
CC(C)(C)OC(=O)NC(CCC(=O)N)C(=O)O
C10H18N2O5
MFCD00065571
boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid

Specifications

White
13726-85-7
MFCD00065571
VVNYDCGZZSTUBC-LURJTMIESA-N
(2S)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
83687
Crystal-Powder at 20°C
Nalpha-(tert-Butoxycarbonyl)-L-glutamine
10g
C10H18N2O5
boc-gln-oh, boc-l-glutamine, boc-glutamine, tert-butoxycarbonyl-l-glutamine, s-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, nalpha-tert-butoxycarbonyl-l-glutamine, n-tert-butoxycarbonyl-l-glutamine, boc-gln, nalpha-boc-l-glutamine, n2-tert-butoxycarbonyl-l-glutamine
CC(C)(C)OC(=O)NC(CCC(=O)N)C(=O)O
246.263
246.26
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : Yes