Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-p-Toluenesulfonyl-L-arginine, 98%

Chemical Identifiers

Chemical Name or Material Nalpha-p-Toluenesulfonyl-L-arginine
CAS 1159-15-5
Formula Weight 328.39
MDL Number MFCD00019732
Synonym Tos-Arg-OH; Nα-Tosyl-L-arginine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6328706

 
alfa aesar™
H6328706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6328714

 
alfa aesar™
H6328714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.39
Chemical Name or Material Nalpha-p-Toluenesulfonyl-L-arginine
Physical Form Solid
Assay 98%
MDL Number MFCD00019732
CAS 1159-15-5
Synonym Tos-Arg-OH; Nα-Tosyl-L-arginine
Quantity 5g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C