missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-p-Toluenesulfonyl-L-arginine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6328714
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Nalpha-p-Toluenesulfonyl-L-arginine | |
C13H20N4O4S | |
tos-arg-oh, n-tosyl-l-arginine, n-p-tosyl-l-arginine, tosyl-l-arginine, nalpha-tosyl-l-arginine, nalpha-p-toluenesulfonyl-l-arginine, p-tosyl-l-arginine, l-arginine, n2-4-methylphenyl sulfonyl, 2s-5-carbamimidamido-2-4-methylbenzenesulfonamido pentanoic acid, tosyl arginine | |
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O | |
328.39 | |
328.39 | |
25 g |
1159-15-5 | |
MFCD00019732 | |
KFNRNFXZFIRNEO-UHFFFAOYNA-N | |
(2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoic acid | |
52501 | |
98% | |
Solid |
Chemical Identifiers
1159-15-5 | |
328.39 | |
KFNRNFXZFIRNEO-UHFFFAOYNA-N | |
52501 | |
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(O)=O |
C13H20N4O4S | |
MFCD00019732 | |
tos-arg-oh, n-tosyl-l-arginine, n-p-tosyl-l-arginine, tosyl-l-arginine, nalpha-tosyl-l-arginine, nalpha-p-toluenesulfonyl-l-arginine, p-tosyl-l-arginine, l-arginine, n2-4-methylphenyl sulfonyl, 2s-5-carbamimidamido-2-4-methylbenzenesulfonamido pentanoic acid, tosyl arginine | |
(2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold