missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%

Inhibitor of papain and trypsin

$107.49 - $436.42

Chemical Identifiers

CAS 4238-41-9
Molecular Formula C14H22Cl2N2O3S
Molecular Weight (g/mol) 369.301
MDL Number MFCD00012513
InChI Key YFCUZWYIPBUQBD-UHFFFAOYSA-N
Synonym 1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
PubChem CID 23335676
IUPAC Name N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCCN)C(=O)CCl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63959MF View Documents Alfa Aesar™
J63959MF
50mg Each for $107.49
 
AAJ63959MC View Documents Alfa Aesar™
J63959MC
100mg Each for $183.51
 
AAJ63959MD View Documents Alfa Aesar™
J63959MD
250mg Each for $436.42
 
Specifications

Chemical Identifiers

4238-41-9
369.301
YFCUZWYIPBUQBD-UHFFFAOYSA-N
23335676
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCCN)C(=O)CCl.Cl
C14H22Cl2N2O3S
MFCD00012513
1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride

Specifications

4238-41-9
MFCD00012513
Soluble in water
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCCN)C(=O)CCl.Cl
369.301
369.31
98%
50mg
C14H22Cl2N2O3S
1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride, p-toluenesulfonamide, hydrochloride 7ci, 8ci, nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride
YFCUZWYIPBUQBD-UHFFFAOYSA-N
N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride
23335676
Powder
White
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XT5160000

Recommended Storage : −30° to −10°C