Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha,Nepsilon-Di-Fmoc-D-lysine, 98%

Chemical Identifiers

Chemical Name or Material Nalpha,Nepsilon-Di-Fmoc-D-lysine
CAS 75932-02-4
Formula Weight 590.68
MDL Number MFCD00270743
Synonym Fmoc-D-Lys(Fmoc)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6513103

 
alfa aesar™
H6513103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6513106

 
alfa aesar™
H6513106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65131MD

 
alfa aesar™
H65131MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 590.68
CAS 75932-02-4
Chemical Name or Material Nalpha,Nepsilon-Di-Fmoc-D-lysine
Physical Form Solid
Assay 98%
MDL Number MFCD00270743
Synonym Fmoc-D-Lys(Fmoc)-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C