Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt
CAS 15098-69-8
Formula Weight 527.74
MDL Number MFCD00038892
Beilstein 6262378
Synonym Boc-Lys(Boc)-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6278514

  SDS
alfa aesar™
H6278514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6278522

  SDS
alfa aesar™
H6278522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 527.74
Melting Point 139° to 143°C
Assay 98%
Synonym Boc-Lys(Boc)-OH.DCHA
MDL Number MFCD00038892
Physical Form Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt
Beilstein 6262378
Quantity 25g
CAS 15098-69-8
Storage Note 1 -30° to -10°C
Color White
Safety and Handling

-30° to -10°C