missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha,Nepsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, TCI America™

$32.08 - $97.78

Chemical Identifiers

CAS 78081-87-5
Molecular Formula C36H34N2O6
Molecular Weight (g/mol) 590.676
MDL Number MFCD00080275
InChI Key BMJRTKDVFXYEFS-XIFFEERXSA-N
Synonym fmoc-lys fmoc-oh, n-a,n-epsilon-di-fmoc-l-lysine, nalpha,nepsilon-di-fmoc-l-lysine, n,n'-bis-fmoc-l-lysine, fmoc-l-lys fmoc-oh, n, a,n, a-di-fmoc-l-lysine, 2s-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoclys fmoc oh, pubchem18973, n, n'-bis-fmoc-l-lysine
PubChem CID 13783708
IUPAC Name (2S)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49315G View Documents TCI America
B49315G
5g Each for $32.08
 
B493125G View Documents TCI America
B493125G
25g Each for $97.78
 
Specifications

Chemical Identifiers

78081-87-5
590.676
BMJRTKDVFXYEFS-XIFFEERXSA-N
13783708
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
C36H34N2O6
MFCD00080275
fmoc-lys fmoc-oh, n-a,n-epsilon-di-fmoc-l-lysine, nalpha,nepsilon-di-fmoc-l-lysine, n,n'-bis-fmoc-l-lysine, fmoc-l-lys fmoc-oh, n, a,n, a-di-fmoc-l-lysine, 2s-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoclys fmoc oh, pubchem18973, n, n'-bis-fmoc-l-lysine
(2S)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid

Specifications

White
5g
C36H34N2O6
fmoc-lys fmoc-oh, n-a,n-epsilon-di-fmoc-l-lysine, nalpha,nepsilon-di-fmoc-l-lysine, n,n'-bis-fmoc-l-lysine, fmoc-l-lys fmoc-oh, n, a,n, a-di-fmoc-l-lysine, 2s-2,6-bis 9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoclys fmoc oh, pubchem18973, n, n'-bis-fmoc-l-lysine
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
590.676
590.68
≥98.0% (HPLC,T)
135°C
78081-87-5
MFCD00080275
BMJRTKDVFXYEFS-XIFFEERXSA-N
(2S)-2,6-bis(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
13783708
Crystal-Powder at 20°C
Nalpha,Nepsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator