Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine
CAS 83792-47-6
Formula Weight 550.63
MDL Number MFCD00063331
Beilstein 5198802
Synonym Fmoc-Arg(Tos)-OH; Nα-Fmoc-Nω-tosyl-L-arginine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6635106

 
alfa aesar™
H6635106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6635114

 
alfa aesar™
H6635114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6635122

 
alfa aesar™
H6635122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 550.63
CAS 83792-47-6
Chemical Name or Material Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine
MDL Number MFCD00063331
Synonym Fmoc-Arg(Tos)-OH; Nα-Fmoc-Nω-tosyl-L-arginine
Quantity 5g
Melting Point 80°C
Physical Form Powder
Storage Note 1 2° to 8°C
Beilstein 5198802
Color White
Assay 95%
Safety and Handling

2° to 8°C