missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6635114
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
80°C | |
83792-47-6 | |
C28H30N4O6S | |
5198802 | |
JRRARHJPRLAGNT-VWLOTQADSA-N | |
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid | |
3010412 | |
95% | |
Powder |
Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine | |
White | |
MFCD00063331 | |
fmoc-arg tos-oh, nalpha-fmoc-nomega-tosyl-l-arginine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine, fmoc-arginine tos, fmoc-l-arg tos-oh, arginine derivative 6, pubchem18947, fmoc-arg tos-oh et2o, fmoc-n-gamma-tosyl-l-arginine etherate, fmoc-nw-4-toluenesulfonyl-l-arginine | |
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N | |
550.63 | |
550.63 | |
25 g |
Chemical Identifiers
83792-47-6 | |
550.63 | |
JRRARHJPRLAGNT-VWLOTQADSA-N | |
3010412 | |
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N |
C28H30N4O6S | |
MFCD00063331 | |
fmoc-arg tos-oh, nalpha-fmoc-nomega-tosyl-l-arginine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine, fmoc-arginine tos, fmoc-l-arg tos-oh, arginine derivative 6, pubchem18947, fmoc-arg tos-oh et2o, fmoc-n-gamma-tosyl-l-arginine etherate, fmoc-nw-4-toluenesulfonyl-l-arginine | |
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold