Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nomega-nitro-L-arginine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Nomega-nitro-L-arginine
CAS 58111-94-7
Formula Weight 441.44
MDL Number MFCD00065617
Beilstein 7060334
Synonym Fmoc-Arg(NO2)-OH; (S)-2-(Fmoc-amino)-5-(3-nitroguanidino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6511406

 
alfa aesar™
H6511406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6511414

 
alfa aesar™
H6511414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 441.44
Physical Form Powder
Assay 95%
Synonym Fmoc-Arg(NO2)-OH; (S)-2-(Fmoc-amino)-5-(3-nitroguanidino)pentanoic acid
Chemical Name or Material Nalpha-Fmoc-Nomega-nitro-L-arginine
CAS 58111-94-7
Beilstein 7060334
Color White
Quantity 5g
Storage Note 1 2° to 8°C
MDL Number MFCD00065617
Safety and Handling

2° to 8°C