missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 95%

Manufacturer:  Alfa Aesar™ H6390706

 View more versions of this product

Catalog No. AAH6390706



Specifications

Specifications

Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine
C31H36N4O7S
fmoc-arg mtr-oh, fmoc-l-arg mtr, fmoc-arg mtr, fmoc-l-arg mtr-oh, ambotzfaa1700, fmoc-arginine mtr-oh, fmoc-l-arginine mtr, pubchem10000, fmoc-ng-mtr-l-arginine hemiisopropyl ether, fmoc-arg mtr-oh tlc
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N)C)C)OC
608.71
608.71
95%
5g
98930-01-9
MFCD00065616
LKGHIEITYHYVED-SANMLTNESA-N
(2S)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
3010428
Powder
White
Moisture Sensitive

Chemical Identifiers

98930-01-9
608.71
LKGHIEITYHYVED-SANMLTNESA-N
3010428
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N)C)C)OC
C31H36N4O7S
MFCD00065616
fmoc-arg mtr-oh, fmoc-l-arg mtr, fmoc-arg mtr, fmoc-l-arg mtr-oh, ambotzfaa1700, fmoc-arginine mtr-oh, fmoc-l-arginine mtr, pubchem10000, fmoc-ng-mtr-l-arginine hemiisopropyl ether, fmoc-arg mtr-oh tlc
(2S)-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : -30° to -10°C

SDS Documents