Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine
CAS 120075-24-3
Formula Weight 608.71
MDL Number MFCD00065616
Synonym Fmoc-D-Arg(Mtr)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6619403

 
alfa aesar™
H6619403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6619406

 
alfa aesar™
H6619406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6619414

 
alfa aesar™
H6619414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 608.71
MDL Number MFCD00065616
CAS 120075-24-3
Physical Form Solid
Assay 95%
Synonym Fmoc-D-Arg(Mtr)-OH
Storage Note 1 2° to 8°C
Quantity 1g
Chemical Name or Material Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine
Safety and Handling

2° to 8°C