Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
CAS 76265-69-5
Formula Weight 464.43
MDL Number MFCD00153360
Synonym Fmoc-Lys(Tfa)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6224303

 
alfa aesar™
H6224303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6224306

 
alfa aesar™
H6224306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6224314

 
alfa aesar™
H6224314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 464.43
Melting Point 170° to 174°C
Chemical Name or Material Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
Physical Form Solid
Assay 95%
MDL Number MFCD00153360
CAS 76265-69-5
Synonym Fmoc-Lys(Tfa)-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C