missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6224306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
170°C to 174°C | |
76265-69-5 | |
MFCD00153360 | |
ZVLMWTPNDXNXSZ-TWYLJJHKNA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid | |
16213176 | |
95% | |
Solid |
Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine | |
C23H23F3N2O5 | |
fmoc-lys tfa-oh, n2-9h-fluoren-9-yl methoxy carbonyl-n6-2,2,2-trifluoroacetyl-l-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, fmoc-n-epsilon-trifluoroacetyl-l-lysine, n, a-fmoc-n, a-trifluoroacetyl-l-lysine, a-fmoc-n, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-trifluoroacetyl-l-lysine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-trifluoroacetyl-l-lysine | |
OC(=O)[C@H](CCCCNC(=O)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
464.44 | |
464.43 | |
5 g |
Chemical Identifiers
76265-69-5 | |
464.44 | |
ZVLMWTPNDXNXSZ-TWYLJJHKNA-N | |
16213176 | |
OC(=O)[C@H](CCCCNC(=O)C(F)(F)F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C23H23F3N2O5 | |
MFCD00153360 | |
fmoc-lys tfa-oh, n2-9h-fluoren-9-yl methoxy carbonyl-n6-2,2,2-trifluoroacetyl-l-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, fmoc-n-epsilon-trifluoroacetyl-l-lysine, n, a-fmoc-n, a-trifluoroacetyl-l-lysine, a-fmoc-n, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-trifluoroacetyl-l-lysine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n∼2∼-9h-fluoren-9-yl methoxy carbonyl-n∼6∼-trifluoroacetyl-l-lysine | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold