missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine, 95%

Manufacturer:  Alfa Aesar™ H6224314

 View more versions of this product

Catalog No. AAH6224314



Specifications

Specifications

Nalpha-Fmoc-Nepsilon-trifluoracetyl-L-lysine
C23H23F3N2O5
fmoc-lys tfa-oh, n2-9h-fluoren-9-yl methoxy carbonyl-n6-2,2,2-trifluoroacetyl-l-lysine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, fmoc-n-epsilon-trifluoroacetyl-l-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, fmoc-lys-tfa-oh, n, a-fmoc-n, a-trifluoroacetyl-l-lysine, fmoc-lys tfa-oh hplc, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-trifluoroacetyl-l-lysine
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC(=O)C(F)(F)F)C(=O)O
464.441
464.43
95%
25g
76265-69-5
MFCD00153360
ZVLMWTPNDXNXSZ-IBGZPJMESA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
16213176
Solid
170°C to 174°C

Chemical Identifiers

76265-69-5
464.441
ZVLMWTPNDXNXSZ-IBGZPJMESA-N
16213176
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC(=O)C(F)(F)F)C(=O)O
C23H23F3N2O5
MFCD00153360
fmoc-lys tfa-oh, n2-9h-fluoren-9-yl methoxy carbonyl-n6-2,2,2-trifluoroacetyl-l-lysine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid, fmoc-n-epsilon-trifluoroacetyl-l-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-trifluoracetyl-l-lysine, fmoc-lys-tfa-oh, n, a-fmoc-n, a-trifluoroacetyl-l-lysine, fmoc-lys tfa-oh hplc, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid, n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-trifluoroacetyl-l-lysine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents